N-Ethyl-4-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Ethylsulphamoyl)benzeneboronic acid

BB-2426-5G BB-2426-1G
COBBBB-2426-5GEA 1690 EUR
COBBBB-2426-5G COBBBB-2426-1G
N-Ethyl-4-boronobenzolsulfonamid
N-Ethyl-4-boronobenzolsulfonamid
Formel: C₈H₁₂BNO₄S
Molecular Weight: 229,06 g/mol
MDL: MFCD07363751
CAS-Nummer: 871329-65-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR