N-Methyl-4-boronobenzolsulfonamid
Lieferant: COMBI-BLOCKS
Synonyme:
4-Methylsulphamoylbenzeneboronic acid, (4-(N-Methylsulphamoyl)phenyl)boronic acid
BB-2610-5G
BB-2610-1G
COBBBB-2610-5GEA
1140
EUR
COBBBB-2610-5G
COBBBB-2610-1G
N-Methyl-4-boronobenzolsulfonamid
N-Methyl-4-boronobenzolsulfonamid
Formel:
C₇H₁₀BNO₄S Molecular Weight: 215.03 g/mol |
MDL:
MFCD06659864 CAS-Nummer: 226396-31-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...