N-Cyclohexyl-3-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

BB-2619-5G BB-2619-1G
COBBBB-2619-5GEA 2350 EUR
COBBBB-2619-5G COBBBB-2619-1G
N-Cyclohexyl-3-boronobenzolsulfonamid
N-Cyclohexyl-3-boronobenzolsulfonamid
Formel: C₁₂H₁₈BNO₄S
MDL: MFCD07783859
CAS-Nummer: 871329-79-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR