N-Cyclopropyl-3-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Cyclopropylsulphamoyl)benzeneboronic acid, (3-(N-Cyclopropylsulphamoyl)phenyl)boronic acid

BB-2620-5G BB-2620-1G
COBBBB-2620-5GEA 1490 EUR
COBBBB-2620-5G COBBBB-2620-1G
N-Cyclopropyl-3-boronobenzolsulfonamid
N-Cyclopropyl-3-boronobenzolsulfonamid
Formel: C₉H₁₂BNO₄S
Molecular Weight: 241.07 g/mol
MDL: MFCD08436035
CAS-Nummer: 913835-28-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR