N-Cyclohexyl-4-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

BB-2429-1G BB-2429-5G
COBBBB-2429-1GEA 0 EUR
COBBBB-2429-1G COBBBB-2429-5G
N-Cyclohexyl-4-boronobenzolsulfonamid
N-Cyclohexyl-4-boronobenzolsulfonamid
Formel: C₁₂H₁₈BNO₄S
MDL: MFCD07363754
CAS-Nummer: 871329-66-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR