N-Cyclopropyl-4-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Cyclopropylsulphamoyl)benzeneboronic acid, (4-(N-Cyclopropylsulphamoyl)phenyl)boronic acid

BB-2430-5G BB-2430-1G
COBBBB-2430-5GEA 1410 EUR
COBBBB-2430-5G COBBBB-2430-1G
N-Cyclopropyl-4-boronobenzolsulfonamid
N-Cyclopropyl-4-boronobenzolsulfonamid
Formel: C₉H₁₂BNO₄S
Molecular Weight: 241.07 g/mol
MDL: MFCD07363755
CAS-Nummer: 871329-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR