N-p-Tolyl-4-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-[(4-Methylphenyl)sulphamoyl]benzeneboronic acid

BB-5048-5G BB-5048-1G
COBBBB-5048-5GEA 692 EUR
COBBBB-5048-5G COBBBB-5048-1G
N-p-Tolyl-4-boronobenzolsulfonamid
N-p-Tolyl-4-boronobenzolsulfonamid
Formel: C₁₃H₁₄BNO₄S
MDL: MFCD09878329
CAS-Nummer: 957062-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR