2-Brom-6-chlor-4-(trifluormethoxy)anilin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-6-chloro-4-trifluoromethoxyaniline

AN-1535-5G AN-1535-1G
COBBAN-1535-5GEA 1170 EUR
COBBAN-1535-5G COBBAN-1535-1G
2-Brom-6-chlor-4-(trifluormethoxy)anilin
2-Brom-6-chlor-4-(trifluormethoxy)anilin
Formel: C₇H₄BrClF₃NO
Molecular Weight: 290,47 g/mol
MDL: MFCD16618925
CAS-Nummer: 1228108-74-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR