3-Brom-5-fluoranthranilsäure

Lieferant: Thermo Scientific

Synonyme: 2-Amino-3-bromo-5-fluorobenzoic acid

SEW05302.250 SEW05302.1
MAYBSEW05302.250EA 0 EUR
MAYBSEW05302.250 MAYBSEW05302.1
3-Brom-5-fluoranthranilsäure
3-Brom-5-fluoranthranilsäure
Formel: C₇H₅BrFNO₂
Molecular Weight: 234,02 g/mol
MDL: MFCD00662509
CAS-Nummer: 259269-84-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR