2-Brom-5-methoxyanilin Hydrochlorid

Lieferant: COMBI-BLOCKS

HC-6911-5G HC-6911-1G
COBBHC-6911-5GEA 124 EUR
COBBHC-6911-5G COBBHC-6911-1G
2-Brom-5-methoxyanilin Hydrochlorid
2-Brom-5-methoxyanilin Hydrochlorid
Formel: C₇H₉BrClNO
Molecular Weight: 238,5095 g/mol
MDL: MFCD10699672
CAS-Nummer: 129968-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR