3,4-Dihydroxy-5-methoxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

464120-1G 464120-5G
SIAL464120-1GEA 59.9 EUR
SIAL464120-1G SIAL464120-5G
3,4-Dihydroxy-5-methoxybenzaldehyd, Sigma-Aldrich®
3,4-Dihydroxy-5-methoxybenzaldehyde
Formel: C₈H₈O₄
Molecular Weight: 168.1467 g/mol
MDL: MFCD00016610
CAS-Nummer: 3934-87-0
EINECS: 223-513-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR