3-Chlor-5-hydroxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

PN-5959-1G PN-5959-250M
COBBPN-5959-1GEA 692 EUR
COBBPN-5959-1G COBBPN-5959-250M
3-Chlor-5-hydroxyphenylboronsäurepinakolester
3-Chlor-5-hydroxyphenylboronsäurepinakolester
Formel: C₁₂H₁₆BClO₃
Molecular Weight: 254.5200 g/mol
MDL: MFCD12546581
CAS-Nummer: 960388-56-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR