2-Chlor-5-formylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Chloro-5-formylphenyl)boronic acid

BB-9018-5G BB-9018-1G
COBBBB-9018-5GEA 179 EUR
COBBBB-9018-5G COBBBB-9018-1G
2-Chlor-5-formylphenylboronsäure
2-Chlor-5-formylphenylboronsäure
Formel: C₇H₆BClO₃
Molecular Weight: 184,3847 g/mol
MDL: MFCD06656269
CAS-Nummer: 1150114-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR