4-Chlor-2-formylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chloro-2-formylbenzeneboronic acid

BB-3489-5G BB-3489-1G
COBBBB-3489-5GEA 759 EUR
COBBBB-3489-5G COBBBB-3489-1G
4-Chlor-2-formylphenylboronsäure
4-Chlor-2-formylphenylboronsäure
Formel: C₇H₆BClO₃
Molecular Weight: 184,39 g/mol
MDL: MFCD08689492
CAS-Nummer: 913835-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR