2-Chlor-5-hydroxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Chloro-5-hydroxyphenyl)boronic acid

BB-3478-5G BB-3478-1G
COBBBB-3478-5GEA 0 EUR
COBBBB-3478-5G COBBBB-3478-1G
2-Chlor-5-hydroxyphenylboronsäure
2-Chlor-5-hydroxyphenylboronsäure
Formel: C₆H₆BClO₃
Molecular Weight: 172.3700 g/mol
MDL: MFCD08689478
CAS-Nummer: 913835-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR