3-Chlor-2-hydroxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-2-hydroxyphenyl)boronic acid

BB-8562-1G BB-8562-5G
COBBBB-8562-1GEA 127 EUR
COBBBB-8562-1G COBBBB-8562-5G
3-Chlor-2-hydroxyphenylboronsäure
3-Chlor-2-hydroxyphenylboronsäure
Formel: C₆H₆BClO₃
Molecular Weight: 172,3740 g/mol
MDL: MFCD08689551
CAS-Nummer: 951655-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR