3-Chlor-4-hydroxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Chloro-4-hydroxyphenyl)boronic acid
BB-3482-5G
BB-3482-1G
COBBBB-3482-5GEA
257
EUR
COBBBB-3482-5G
COBBBB-3482-1G
3-Chlor-4-hydroxyphenylboronsäure
3-Chlor-4-hydroxyphenylboronsäure
Formel:
C₆H₆BClO₃ Molecular Weight: 172,3700 g/mol |
MDL:
MFCD09258739 CAS-Nummer: 182344-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...