3-Chlor-5-hydroxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-Chloro-5-hydroxyphenyl)boronic acid

BB-5959-5G BB-5959-1G BB-5959-250M
COBBBB-5959-5GEA 2160 EUR
COBBBB-5959-5G COBBBB-5959-1G COBBBB-5959-250M
3-Chlor-5-hydroxyphenylboronsäure
3-Chlor-5-hydroxyphenylboronsäure
Formel: C₆H₆BClO₃
Molecular Weight: 172,3700 g/mol
MDL: MFCD15143456
CAS-Nummer: 1214900-52-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR