5-Chlor-2-hydroxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Chloro-2-hydroxybenzeneboronic acid

BB-2604-5G BB-2604-1G
COBBBB-2604-5GEA 324 EUR
COBBBB-2604-5G COBBBB-2604-1G
5-Chlor-2-hydroxyphenylboronsäure
5-Chlor-2-hydroxyphenylboronsäure
Formel: C₆H₆BClO₃
Molecular Weight: 172,37 g/mol
MDL: MFCD06659863
CAS-Nummer: 89488-25-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR