4-Chlor-3-hydroxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-Chloro-3-hydroxyphenyl)boronic acid
BB-5718-1G
BB-5718-250M
COBBBB-5718-1GEA
471
EUR
COBBBB-5718-1G
COBBBB-5718-250M
4-Chlor-3-hydroxyphenylboronsäure
4-Chlor-3-hydroxyphenylboronsäure
Formel:
C₆H₆BClO₃ Molecular Weight: 172,3740 g/mol |
MDL:
MFCD10697413 CAS-Nummer: 915201-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...