2-Chlor-3-ethoxy-6-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-5022-5G
BB-5022-1G
COBBBB-5022-5GEA
370
EUR
COBBBB-5022-5G
COBBBB-5022-1G
2-Chlor-3-ethoxy-6-fluorphenylboronsäure
2-Chlor-3-ethoxy-6-fluorphenylboronsäure
Formel:
C₈H₉BClFO₃ Molecular Weight: 218.4200 g/mol |
MDL:
MFCD09800880 CAS-Nummer: 957120-93-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...