2,6-Difluor-4-formylphenylboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2,6-Difluoro-4-formylphenylboronic acid, pinacol ester, 3,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
M-Clarity™ Qualitätsstufe = MQ100
663514-1G
663514-5G
SIAL663514-1GEA
103
EUR
SIAL663514-1G
SIAL663514-5G
2,6-Difluor-4-formylphenylboronsäurepinakolester, Sigma-Aldrich®
2,6-Difluor-4-formylphenylboronsäurepinakolester
Formel:
C₁₃H₁₅BF₂O₃ Molecular Weight: 268,600 g/mol |
MDL:
MFCD08276848 CAS-Nummer: 870717-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...