2,4-Difluor-3-formylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2,4-Difluoro-3-formylphenyl)boronic acid

BB-8002-5G BB-8002-1G
COBBBB-8002-5GEA 227 EUR
COBBBB-8002-5G COBBBB-8002-1G
2,4-Difluor-3-formylphenylboronsäure
2,4-Difluor-3-formylphenylboronsäure
Formel: C₇H₅BF₂O₃
Molecular Weight: 185.9200 g/mol
MDL: MFCD04974116
CAS-Nummer: 870718-06-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR