2,4-Difluor-3-formylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2,4-Difluoro-3-formylphenyl)boronic acid

M-Clarity™ Qualitätsstufe = MQ100

597406-10G 597406-1G
SIAL597406-10GEA 286 EUR
SIAL597406-10G SIAL597406-1G
2,4-Difluor-3-formylphenylboronsäure, Sigma-Aldrich®
2,4-Difluor-3-formylphenylboronsäure
Formel: C₇H₅BF₂O₃
Molecular Weight: 185,9200 g/mol
MDL: MFCD04974116
CAS-Nummer: 870718-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR