2-Fluor-4-formylphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8512-5G BB-8512-1G
COBBBB-8512-5GEA 85.8 EUR
COBBBB-8512-5G COBBBB-8512-1G
2-Fluor-4-formylphenylboronsäure
2-Fluor-4-formylphenylboronsäure
Formel: C₇H₆BFO₃
Molecular Weight: 167,9300 g/mol
MDL: MFCD07367058
CAS-Nummer: 871126-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR