2-Fluor-4-formylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

657344-10G 657344-2G
SIAL657344-10GEA 283 EUR
SIAL657344-10G SIAL657344-2G
2-Fluor-4-formylphenylboronsäure, Sigma-Aldrich®
2-Fluor-4-formylphenylboronsäure
Formel: C₇H₆BFO₃
Molecular Weight: 167,9300 g/mol
MDL: MFCD07367058
CAS-Nummer: 871126-22-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR