2-Fluor-5-formylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-5-formylbenzeneboronic acid

BB-8380-5G BB-8380-25G BB-8380-1G
COBBBB-8380-5GEA 93.4 EUR
COBBBB-8380-5G COBBBB-8380-25G COBBBB-8380-1G
2-Fluor-5-formylphenylboronsäure
2-Fluor-5-formylphenylboronsäure
Formel: C₇H₆BFO₃
Molecular Weight: 167,93 g/mol
MDL: MFCD03095100
CAS-Nummer: 352534-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR