2-Fluor-5-formylphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Fluoro-5-formylbenzeneboronic acid

016205-1G 016205-5G
FLUO016205-1GEA 116 EUR
FLUO016205-1G FLUO016205-5G
2-Fluor-5-formylphenylboronsäure
2-Fluor-5-formylphenylboronsäure
Formel: C₇H₆BFO₃
Molecular Weight: 167.93 g/mol
MDL: MFCD03095100
CAS-Nummer: 352534-79-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR