3,5-Difluor-4-formylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3,5-Difluoro-4-formylphenyl)boronic acid

M-Clarity™ Qualitätsstufe = MQ100

635782-1G 635782-5G
SIAL635782-1GEA 152 EUR
SIAL635782-1G SIAL635782-5G
3,5-Difluor-4-formylphenylboronsäure, Sigma-Aldrich®
3,5-Difluor-4-formylphenylboronsäure
Formel: C₇H₅BF₂O₃
Molecular Weight: 185,9206 g/mol
MDL: MFCD07784344
CAS-Nummer: 870718-11-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR