4-Fluor-3-formylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-3-formylbenzeneboronic acid, (4-Fluoro-3-formylphenyl)boronic acid

BB-3831-5G BB-3831-25G BB-3831-1G
COBBBB-3831-5GEA 127 EUR
COBBBB-3831-5G COBBBB-3831-25G COBBBB-3831-1G
4-Fluor-3-formylphenylboronsäure
4-Fluor-3-formylphenylboronsäure
Formel: C₇H₆BFO₃
Molecular Weight: 167.93 g/mol
MDL: MFCD02093074
CAS-Nummer: 374538-01-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR