4-Fluor-3-formylphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluoro-3-formylbenzeneboronic acid, (4-Fluoro-3-formylphenyl)boronic acid

M-Clarity™ Qualitätsstufe = MQ100

683825-1G 683825-5G
SIAL683825-1GEA 39 EUR
SIAL683825-1G SIAL683825-5G
4-Fluor-3-formylphenylboronsäure, Sigma-Aldrich®
4-Fluor-3-formylphenylboronsäure
Formel: C₇H₆BFO₃
Molecular Weight: 167,93 g/mol
MDL: MFCD02093074
CAS-Nummer: 374538-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR