N-(4-(Trifluormethoxy)phenyl)-4-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-{[4-(Trifluoromethoxy)phenyl]sulphamoyl}benzeneboronic acid

BB-5051-5G BB-5051-1G
COBBBB-5051-5GEA 0 EUR
COBBBB-5051-5G COBBBB-5051-1G
N-(4-(Trifluormethoxy)phenyl)-4-boronobenzolsulfonamid
N-(4-(Trifluormethoxy)phenyl)-4-boronobenzolsulfonamid
Formel: C₁₃H₁₁BF₃NO₅S
MDL: MFCD09878331
CAS-Nummer: 957062-98-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR