N-(4-Fluorphenyl)-4-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-[N-(4-Fluorophenyl)sulphamoyl]benzeneboronic acid, (4-(N-(4-Fluorophenyl)sulphamoyl)phenyl)boronic acid

BB-5050-5G BB-5050-1G
COBBBB-5050-5GEA 884 EUR
COBBBB-5050-5G COBBBB-5050-1G
N-(4-Fluorphenyl)-4-boronobenzolsulfonamid
N-(4-Fluorphenyl)-4-boronobenzolsulfonamid
Formel: C₁₂H₁₁BFNO₄S
Molecular Weight: 295,09 g/mol
MDL: MFCD09800889
CAS-Nummer: 957121-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR