3-Ethoxy-5-formylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Ethoxy-5-formylphenylboronic acid
BB-8239-1G
BB-8239-250M
COBBBB-8239-1GEA
363
EUR
COBBBB-8239-1G
COBBBB-8239-250M
3-Ethoxy-5-formylphenylboronsäure
3-Ethoxy-5-formylphenylboronsäure
Formel:
C₉H₁₁BO₄ Molecular Weight: 193.992 g/mol |
MDL:
MFCD09038413 CAS-Nummer: 1072952-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...