N,N-Diethyl-3-borono-4-methylbenzolsulfonamid
Lieferant: COMBI-BLOCKS
Synonyme:
(5-(N,N-Diethylsulphamoyl)-2-methylphenyl)boronic acid, 5-Diethylsulphamoyl-2-methylbenzeneboronic acid
BB-2698-5G
BB-2698-1G
COBBBB-2698-5GEA
1580
EUR
COBBBB-2698-5G
COBBBB-2698-1G
N,N-Diethyl-3-borono-4-methylbenzolsulfonamid
N,N-Diethyl-3-borono-4-methylbenzolsulfonamid
Formel:
C₁₁H₁₈BNO₄S Molecular Weight: 271,14 g/mol |
MDL:
MFCD08056356 CAS-Nummer: 871329-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...