N-tert-Butyl-3-borono-4-methylbenzolsulfonamid
Lieferant: COMBI-BLOCKS
Synonyme:
5-tert-Butylsulphamoyl-2-methylbenzeneboronic acid
BB-2900-1G
BB-2900-250M
COBBBB-2900-1GEA
1700
EUR
COBBBB-2900-1G
COBBBB-2900-250M
N-tert-Butyl-3-borono-4-methylbenzolsulfonamid
N-tert-Butyl-3-borono-4-methylbenzolsulfonamid
Formel:
C₁₁H₁₈BNO₄S Molecular Weight: 271,14 g/mol |
MDL:
MFCD08235063 CAS-Nummer: 874219-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...