4'-(2-Chlor-4-fluorphenoxy)acetophenon

Lieferant: FLUOROCHEM

023245-25G 023245-5G
FLUO023245-25GEA 612 EUR
FLUO023245-25G FLUO023245-5G
4'-(2-Chlor-4-fluorphenoxy)acetophenon
4'-(2-Chlor-4-fluorphenoxy)acetophenon
Formel: C₁₄H₁₀BrFO₂
MDL: MFCD06411654
CAS-Nummer: 845866-49-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR