p-Toluolsulfonylacetonitril ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
[(4-Methylphenyl)sulphonyl]acetonitrile
A12857.06
A12857.14
A12857.22
A12857.06EA
25.6
EUR
A12857.06
A12857.14
A12857.22
p-Toluolsulfonylacetonitril ≥98%
p-Toluolsulfonylacetonitril
Purity: 98+%
CAS No.: 5697-44-9
Molecular Formula: C9H9NO2S
Formula Weight: 195.24
Melting Point: 147-152°
Beilstein Reference: 1842131
UN No.: 3439
MDL No.: MFCD00039487
Formel:
C₉H₉NO₂S Molecular Weight: 195,24 g/mol Schmelzpunkt: 147…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039487 CAS-Nummer: 5697-44-9 EINECS: 227-173-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...