4'-(Methylsulfonyl)acetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4'-(Methylsulphonyl)acetophenone

M-Clarity™ Qualitätsstufe = MQ200

549304-25G 549304-5G
SIAL549304-25GEA 99.5 EUR
SIAL549304-25G SIAL549304-5G
4'-(Methylsulfonyl)acetophenon, Sigma-Aldrich®
4'-(Methylsulfonyl)acetophenon
Formel: C₉H₁₀O₃S
Molecular Weight: 198,24 g/mol
MDL: MFCD00025072
CAS-Nummer: 10297-73-1
EINECS: 233-672-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR