2-Hydroxy-3-(trifluormethyl)benzaldehyd ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(Trifluoromethyl)salicylaldehyde
H32858.MD
H32858.03
H32858.MDEA
82.6
EUR
H32858.MD
H32858.03
2-Hydroxy-3-(trifluormethyl)benzaldehyd ≥98%
2-Hydroxy-3-(trifluoromethyl)benzaldehyde
Purity: 98+%
CAS No.: 336628-67-2
Molecular Formula: C8H5F3O2
Formula Weight: 190.12
Melting Point: 54-59°
MDL No.: MFCD03788529
Formel:
C₈H₅F₃O₂ Molecular Weight: 190,12 g/mol Schmelzpunkt: 54…59 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788529 CAS-Nummer: 336628-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...