4'-(2,4-Difluorphenoxy)acetophenon

Lieferant: FLUOROCHEM

023233-25G 023233-5G
FLUO023233-25GEA 1030 EUR
FLUO023233-25G FLUO023233-5G
4'-(2,4-Difluorphenoxy)acetophenon
4'-(2,4-Difluorphenoxy)acetophenon
Formel: C₁₄H₁₀F₂O₂
MDL: MFCD06411650
CAS-Nummer: 845823-27-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR