4'-(2,5-Difluorphenoxy)acetophenon

Lieferant: FLUOROCHEM

023919-25G 023919-5G
FLUO023919-25GEA 648 EUR
FLUO023919-25G FLUO023919-5G
4'-(2,5-Difluorphenoxy)acetophenon
4'-(2,5-Difluorphenoxy)acetophenon
Formel: C₁₄H₁₀F₂O₂
MDL: MFCD08461876
CAS-Nummer: 886497-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR