N-Benzylidenanilin 99

Lieferant: Thermo Fisher Scientific
Achtung

A14746.14 A14746.22 A14746.06
A14746.14EA 143 EUR
A14746.14 A14746.22 A14746.06
N-Benzylidenanilin 99
N-Benzylidenanilin

Purity: 99% CAS No.: 538-51-2 Molecular Formula: C13H11N Formula Weight: 181.24 Flash Point: 110°(230°F) Melting Point: 52-54° MDL No.: MFCD00003027

Formel: C₁₃H₁₁N
Molecular Weight: 181.24 g/mol
Schmelzpunkt: 52…54 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003027
CAS-Nummer: 538-51-2
EINECS: 208-694-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR