4'-(3-Fluorbenzyloxy)acetophenon

Lieferant: FLUOROCHEM

022128-5G 022128-25G
FLUO022128-5GEA 137 EUR
FLUO022128-5G FLUO022128-25G
4'-(3-Fluorbenzyloxy)acetophenon
4'-(3-Fluorbenzyloxy)acetophenon
Formel: C₁₅H₁₃FO₂
MDL: MFCD06411642
CAS-Nummer: 1044063-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR