4'-(4-Fluorbenzyloxy)acetophenon

Lieferant: FLUOROCHEM

008324-1G 008324-5G 008324-25G
FLUO008324-1GEA 31.1 EUR
FLUO008324-1G FLUO008324-5G FLUO008324-25G
4'-(4-Fluorbenzyloxy)acetophenon
4'-(4-Fluorbenzyloxy)acetophenon
Formel: C₁₅H₁₃FO₂
MDL: MFCD00173651
CAS-Nummer: 72293-96-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR