4-Methylbenzoesäureanhydrid ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-methylbenzene-1-carboxylic anhydride
H33951.03
H33951.06
H33951.03EA
35.3
EUR
H33951.03
H33951.06
4-Methylbenzoesäureanhydrid ≥97%
4-Methylbenzoic anhydride
Purity: 97%
CAS No.: 13222-85-0
Molecular Formula: C16H14O3
Formula Weight: 254.29
Melting Point: 86-89°
MDL No.: MFCD00048100
Formel:
C₁₆H₁₄O₃ Molecular Weight: 254,29 g/mol Schmelzpunkt: 86…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00048100 CAS-Nummer: 13222-85-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...