4-Methylbenzoesäureanhydrid
Lieferant: Thermo Scientific
Synonyme:
4-methylbenzene-1-carboxylic anhydride
BTB15092.250
BTB15092.1
MAYBBTB15092.250EA
0
EUR
MAYBBTB15092.250
MAYBBTB15092.1
4-Methylbenzoesäureanhydrid
4-Methylbenzoic anhydride
Formel:
C₁₆H₁₄O₃ Molecular Weight: 254,29 g/mol |
MDL:
MFCD00048100 CAS-Nummer: 13222-85-0 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...