5'-Chlor-2'-hydroxy-4'-methylacetophenon

Lieferant: CHEMPUR

Synonyme: 1-(5-Chloro-2-hydroxy-4-methylphenyl)ethanone

FL12071.POR 012071-10G 012071-25G
CHMPFL12071.POREA 0 EUR
CHMPFL12071.POR FLUO012071-10G FLUO012071-25G
5'-Chlor-2'-hydroxy-4'-methylacetophenon
5'-Chlor-2'-hydroxy-4'-methylacetophenon
Formel: C₉H₉ClO₂
Molecular Weight: 184.6200 g/mol
MDL: MFCD00191920
CAS-Nummer: 28480-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR