4'-Methyl-3'-nitroacetophenon ≥99%
Lieferant: Thermo Scientific
Synonyme: 1-(4-Methyl-3-nitrophenyl)ethanone
L09372.06 L09372.14
L09372.06EA 40.5 EUR
L09372.06 L09372.14
4'-Methyl-3'-nitroacetophenon ≥99%
4'-Methyl-3'-nitroacetophenon
Purity: 99% CAS No.: 5333-27-7 Molecular Formula: C9H9NO3 Formula Weight: 179.18 Melting Point: 58-62° Beilstein Reference: 1953352
Formel: C₉H₉NO₃
Molecular Weight: 179,18 g/mol
Schmelzpunkt: 58…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00033858
CAS-Nummer: 5333-27-7