3-Brom-2-(4-morpholino)pyridin

Lieferant: COMBI-BLOCKS

PY-1240-5G PY-1240-25G
COBBPY-1240-5GEA 98.3 EUR
COBBPY-1240-5G COBBPY-1240-25G
3-Brom-2-(4-morpholino)pyridin
3-Brom-2-(4-morpholino)pyridin
Formel: C₉H₁₁BrN₂O
MDL: MFCD00498942
CAS-Nummer: 54231-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR